missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymidine, 99%

Product Code. 11420403
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420403 100 g 100g
11400403 5 g 5g
11410403 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420403 Supplier Thermo Scientific Chemicals Supplier No. A11493.22
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 50-89-5 | C10H14N2O5 | 242.23 g/mol

Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

Solubility
Soluble in water, methanol, hot ethanol, hot acetone, hot ethyl acetate, pyridine, glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
MDL Number MFCD00006537
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Melting Point 186°C to 190°C
Color White
Odor Odorless
Quantity 100 g
Beilstein 89285
Merck Index 14,9397
Solubility Information Soluble in water,methanol,hot ethanol,hot acetone,hot ethyl acetate,pyridine,glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .
Formula Weight 242.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Thymidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.