missing translation for 'onlineSavingsMsg'
Learn More

Thymidine, MedChemExpress

Product Code. 16402569
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16402569 Supplier Medchem Express Supplier No. HYN1150
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thymidine, a specific precursor of deoxyribonucleic acid, is used as a cell synchronizing agent. Thymidine is a DNA synthesis inhibitor that can arrest cell at G1/S boundary, prior to DNA replication.

Thymidine, a specific precursor of deoxyribonucleic acid, is used as a cell synchronizing agent. Thymidine is a DNA synthesis inhibitor that can arrest cell at G1/S boundary, prior to DNA replication.

Specifications

Chemical Name or Material Thymidine
CAS 50-89-5
Color White
Purity Grade Notes Research
Molecular Formula C10H14N2O5
Quantity 1 g
Synonym DThyd NSC 21548
Solubility Information DMSO : 50 mg/mL (206.42 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (137.60 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(C)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 242.23
Formula Weight 242.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.