missing translation for 'onlineSavingsMsg'
Learn More

Thymidine, MP Biomedicals™

Product Code. 11492051 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492051 25 g 25g
11462051 1 g 1g
11472051 5 g 5g
11482051 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11492051 Supplier MP Biomedicals Supplier No. 0219475425
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in forming hybrid cell lines where cells are incubated in HAT medium. Thymidine, MP Biomedicals, a cell culture reagent, is a nucleoside composed of deoxyribose (a pentose sugar) joined to the pyrimidine base thymine. It is highly stable and appears

Mainly used in forming hybrid cell lines using HAT (hypoxanthine-aminopterin-thymidine) medium to give rise to monoclonal antibodies. Also used in HT and HAT cocktails for hybridoma fusion, selection and cloning.

Deoxythymidine can be phosphorylated with one, two or three phosphoric acid groups, creating respectively dTMP, dTDP or dTTP (deoxythymidine mono-, di-, or triphosphate). The stability of deoxythymidine under standard temperature and pressure (STP) is very high. Thymidine is listed as a chemical teratogen.

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Melting Point 185°C to 191°C
Quantity 25 g
Solubility Information Soluble in water, methanol, hot alcohol, hot acetone, hot ethyl acetate pyridine, glacial acetic acid and HCl (20mg/mL, clear, colorless solution); sparingly soluble in hot chloroform.
Optical Rotation 17.5–19.5° (c=1, water)
Formula Weight 242.2
Percent Purity >99.9%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.