missing translation for 'onlineSavingsMsg'
Learn More

Thymine, MedChemExpress

Product Code. 16433429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16433429 Supplier Medchem Express Supplier No. HYW010450

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM.

Thymine is one of the four nucleobases in the nucleic acid of DNA and can be a target for actions of 5-fluorouracil (5-FU) in cancer treatment, with a Km of 2.3 μM.

Specifications

Chemical Name or Material Thymine
CAS 65-71-4
Color White
Purity Grade Notes Research
Molecular Formula C5H6N2O2
Quantity 1 g
Solubility Information DMSO : 100 mg/mL (792.96 mM; Need ultrasonic) ∣H2O : 5 mg/mL (39.65 mM; Need ultrasonic)
SMILES O=C1NC(C(C)=CN1)=O
Molecular Weight (g/mol) 126.11
Formula Weight 126.11
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.