missing translation for 'onlineSavingsMsg'
Learn More

Thymol, 98+%

Product Code. 11471948
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471948 100 g 100g
11483920 250 g 250g
11481948 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471948 Supplier Thermo Scientific Alfa Aesar Supplier No. A14563.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 89-83-8 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Thymol is used as a preservative in halothane. It acts as an anesthetic, antiseptic in mouth wash, stabilizer in pharmaceutical preparations. It inhibits the growth and lactate production as well as reduces the uptake of cellular glucose within the bacteria. It is an active ingredient in toothpastes like euthymol. It is involved to control varroa mites and prevent fermentation and the growth of mold in bee colonies.

Solubility
Slightly soluble in water. Soluble in chloroform, alcohol and ether.

Notes
Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-83-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002309
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
Synonym 2-Isopropyl-5-methylphenol
PubChem CID 6989
ChEBI CHEBI:27607
IUPAC Name 5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1)C(C)C)O

Specifications

Melting Point 49°C to 52°C
Boiling Point 231°C to 233°C
UN Number UN2430
Beilstein 1907135
Merck Index 14,9399
Solubility Information Slightly soluble in water. Soluble in chloroform,alcohol and ether.
Formula Weight 150.22
Percent Purity ≥98%
Density 0.97
pH 5 to 7
Quantity 100 g
Flash Point 102°C (215°F)
Chemical Name or Material Thymol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thymol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Thymol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.