missing translation for 'onlineSavingsMsg'
Learn More

Thymol, 99%

Product Code. 10616932 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10616932 500 g Plastic bottle 500g
10572931 100 g Plastic bottle 100g
10484025 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10616932 Supplier Thermo Scientific Acros Supplier No. 150335000
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 89-83-8 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-83-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002309
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
Synonym thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
PubChem CID 6989
ChEBI CHEBI:27607
IUPAC Name 5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1)C(C)C)O

Specifications

Melting Point 48.0°C to 52.0°C
Color White
Density 0.9600g/mL
Boiling Point 232.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 06, 532
Residue after Evaporation 0.05% max.
Merck Index 15, 9554
Specific Gravity 0.96
Solubility Information Solubility in water: 0.1% (20°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol, carbon disulfide, chloroform, and ether, slightly soluble in glycerol
Viscosity 4.3 mm2/s (50°C)
Formula Weight 150.22
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Thymol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thymol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 5-methyl-2-(1-methylethyl)-phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.