missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue, ACS reagent

Product Code. 10697401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697401 10 g Glass bottle 10g
10331311 5 g Glass bottle 5g
10288903 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10697401 Supplier Thermo Scientific Chemicals Supplier No. 420930100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76-61-9 | C27H30O5S | 466.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
Synonym TB, Thymolsulfonephthalein
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Clarity Passes Test
Merck Index 15, 9555
Infrared Spectrum Authentic
Formula Weight 466.58
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval Acid range from pH 1.2 (pink) to pH 2.8 (yellow), Alkaline range from pH 8.0 (yellow) to pH 9.2 (blue)
Color Brown to Green
Quantity 10 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Thymol Blue, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.