missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymol Blue, ACS

Product Code. 11377168
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377168 2 g 2g
11367168 10 g 10g
11357168 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377168 Supplier Thermo Scientific Chemicals Supplier No. 016272.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

pH indicator: 1.2 (red) to 2.8 (yellow) to 9.6 (blue) | CAS: 76-61-9 | C27H30O5S | 466.592 g/mol

pH indicator: 1.2 (pink) to 2.8 (yellow) to 9.6 (blue). Thymol Blue is useful as a pH indicator. It is a component of universal indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-61-9
Molecular Formula C27H30O5S
Molecular Weight (g/mol) 466.592
MDL Number MFCD00005869
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
Synonym Thymolsulfonphthalein
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 223°C (decomposition)
Merck Index 14,9400
Solubility Information Soluble in alcohol and dilute alkali solutions; Insoluble in water .
Quantity 2 g
Physical Form Crystalline
Formula Weight 466.6
Grade ACS
Chemical Name or Material Thymol Blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.