missing translation for 'onlineSavingsMsg'
Learn More

Thymol, Extra Pure, SLR, Fisher Chemical

Product Code. 10599580 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10599580 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10599580 Supplier Fisher Chemical Supplier No. T/1450/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 89-83-8
Melting Point 48°C
Color White
pH 5 to 7
Boiling Point 232°C
Packaging Plastic powder jar
Molecular Formula C10H14O
MDL Number 2309
Quantity 100 g
Synonym thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)C(C)C)O
IUPAC Name 5-methyl-2-propan-2-ylphenol
Molecular Weight (g/mol) 150.221
PubChem CID 6989
ChEBI CHEBI:27607
Formula Weight 150.22g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thymol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.