missing translation for 'onlineSavingsMsg'
Learn More

Thymol, TRC

Product Code. 30132608
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132608 50 g 50g
30012862 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132608 Supplier Toronto Research Chemicals Supplier No. TRCT41300050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-83-8
Molecular Formula C10 H14 O
Molecular Weight (g/mol) 150.22
Synonym Thymol, 5-Methyl-2-(methylethyl)phenol, Phenol, 5-methyl-2-(1-methylethyl)-, Thymol (8CI), 5-Methyl-2-(1-methylethyl)phenol, 1-Methyl-3-hydroxy-4-isopropylbenzene, 2-Hydroxy-1-isopropyl-4-methylbenzene, 2-Isopropyl-5-methylphenol, 3-Hydroxy-p-cymene, 3-Methyl-6-isopropylphenol, 5-Methyl-2-isopropyl-1-phenol, 5-Methyl-2-isopropylphenol, 5-Methyl-2-propan-2-ylphenol, 6-Isopropyl-3-methylphenol, 6-Isopropyl-m-cresol, Apiguard, NSC 11215, NSC 47821, NSC 49142, T 0501, Thyme camphor, Thymol Chrystals, m-Thymol, p-Cymen-3-ol
IUPAC Name 5-methyl-2-propan-2-ylphenol
SMILES CC(C)c1ccc(C)cc1O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3
Formula Weight 150.1045
Chemical Name or Material Thymol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.