missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalein, 0.05% w/v solution in ethanol

Product Code. 11440161
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440161 500 mL 500mL
11348977 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440161 Supplier Thermo Scientific Chemicals Supplier No. 038706.AP
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 125-20-2 | C28H30O4 | 430.544 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.544
MDL Number MFCD00005909
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Beilstein 359413
Formula Weight 430.55
Vapor Pressure 57 hPa (150mm Hg) at 20°C
Density 0.92 g/cm3 at 20°C
Color Colorless
Quantity 500 mL
Flash Point 11°C (52°F)
Physical Form Liquid
Chemical Name or Material Thymolphthalein, 0.05% w/v solution in ethanol

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.