missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalein

Product Code. 11466420
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466420 10 g 10g
11476420 50 g 50g
11486420 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466420 Supplier Thermo Scientific Chemicals Supplier No. B23896.09
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 125-20-2 | C28H30O4 | 430.544 g/mol

Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymolphthalein acts as pH indicator, as reagent for blood after decolorizing the alkaline solution by boiling with zinc dust.

Solubility
Soluble in alcohol, acetone and also soluble in dilute alkalis. insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.544
MDL Number MFCD00005909
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
Synonym thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 251°C to 253°C
Quantity 10 g
Beilstein 359413
Merck Index 14,9401
Solubility Information Soluble in alcohol,acetone and also soluble in dilute alkalis. insoluble in water.
Formula Weight 430.55
Chemical Name or Material Thymolphthalein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.