missing translation for 'onlineSavingsMsg'
Learn More

Thymolphthalein, Pure, Solid, Indicator Grade, Fisher Chemical™

Product Code. 10766111 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10766111 25 g 25g
10696652 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10766111 Supplier Fisher Chemical Supplier No. T/1480/46
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 125-20-2
Molecular Formula C28H30O4
Molecular Weight (g/mol) 430.544
MDL Number 5909
InChI Key LDKDGDIWEUUXSH-UHFFFAOYSA-N
PubChem CID 31316
IUPAC Name 3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

Melting Point 251°C
Formula Weight 430.53g/mol
Packaging Glass Jar
Color White
Quantity 25 g
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.