missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Thymolphthalexone

Product Code. 11303106
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303106 1 g 1g
11313106 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303106 Supplier Thermo Scientific Chemicals Supplier No. L10962.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1913-93-5 | C38H44N2O12 | 720.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1913-93-5
Molecular Formula C38H44N2O12
Molecular Weight (g/mol) 720.77
MDL Number MFCD00001619
InChI Key AGXCADFSYJNMEG-UHFFFAOYNA-N
Synonym thymolphthalexone, thymolphthalein complexon, thymolphthalein complexone, thymolphthalexon, thymolphthalein-3,3'-bismethyliminodiacetic acid, 3-1-3-bis carboxymethyl amino methyl-4-hydroxy-5-isopropyl-2-methylphenyl-3-oxo-2-benzofuran-1-yl-6-hydroxy-5-isopropyl-2-methylphenyl methyl carboxymethyl amino acetic acid, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-2-methyl-5-1-methylethyl-3,1-phenylene methylene bis n-carboxymethyl glycine, thymophthalexon, 3',3-bis n,n-di carboxymethyl aminomethyl thymolphthalein, 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene methylenenitrilo tetraacetic acid
PubChem CID 112918
IUPAC Name 2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid
SMILES CC(C)C1=CC(=C(C)C(C[NH+](CC(O)=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(C[NH+](CC(O)=O)CC([O-])=O)=C1C

Spezifikation

Quantity 1 g
Beilstein 1339087
Formula Weight 720.78
Chemical Name or Material Thymolphthalexone

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.