missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thyroxine (T4) Competitive ELISA Kit

Product Code. 15845863
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15845863 96 Tests Pack of 96
17865217 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15845863 Supplier Invitrogen™ Supplier No. EIAT4C
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ELISA

The Thyroxine (T4) ELISA quantitates Thyroxine (T4) in serum, plasma, urine, dried fecal extracts, or cell culture medium. The assay will exclusively recognize both natural and recombinant Thyroxine (T4). Principle of the method The Thyroxine (T4) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Thyroxine is the main hormone produced by the thyroid gland. The thyroid hormones, triiodothyronine (T3) and thyroxine (T4), are tyrosine-based hormones produced by the thyroid gland that are primarily responsible for regulation of metabolism. Iodine is necessary for the production of T3 and T4. The major form of thyroid hormone in the blood is thyroxine, which has a longer half-life than T3. T4 is converted to the active T3 within cells by deiodinases (5'-iodinase). These are further processed by decarboxylation and deiodination to produce iodothyronamine and thyronamine. Thyroid hormone production is regulated via pituitary thyrotropin (TSH) modulation of thyroxine prohormone secretion by the thyroid gland and regulation of active triiodothyronine production. Thyroxine is identical across all species.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.625-20 ng/mL
Assay Sensitivity 0.33 ng/mL
Content And Storage Pre-coated plate(s), standard, detector antibody, HRP conjugate, assay buffer, wash buffer, chromogen, stop solution, plate covers, and lot-specific technical data sheet.

Store at –20°C.
Description Thyroxine (T4) Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Thyroid hormone
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 96 assays
Target Species Chemical
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Absorbance Microplate Reader
Interassay CV 7%
Intraassay CV 3%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases C15H11I4NO4
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Metabolism
Sample Volume Fecal Extract 10 μL, Plasma 1 μL, Serum 1 μL, Supernatant 10 μL, Urine 2.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Thyroxine (T4)
Test Time 1 hr 20 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.