missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiagabine hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Tiagabine hydrochloride is a potent and selective GABA reuptake inhibitor, used as an anticonvulsant agent, with IC50s of 67, 446 and 182 nM for [3H]GABA uptake in Synaptosomes, Neurons and Glia, respectively.
Specifications
Specifications
| Chemical Name or Material | Tiagabine hydrochloride |
| CAS | 145821-59-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H26ClNO2S2 |
| Quantity | 100 mg |
| Synonym | NO050328 hydrochloride NO328 hydrochloride TGB hydrochloride |
| Solubility Information | H2O : 100 mg/mL (242.71 mM; Need ultrasonic) ∣DMSO : ≥ 53 mg/mL (128.64 mM) |
| SMILES | O=C([C@H]1CN(CC/C=C(C2=C(C)C=CS2)/C3=C(C)C=CS3)CCC1)O.[H]Cl |
| Molecular Weight (g/mol) | 412.01 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction