missing translation for 'onlineSavingsMsg'
Learn More

Ticarcillin Disodium Salt (Contain 10% Methanol), TRC

Product Code. 30455551
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455551 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455551 Supplier Toronto Research Chemicals Supplier No. TRCT43790150MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ticarcillin Disodium Salt
CAS 29457-07-6
Molecular Formula C15 H14 N2 O6 S2 . 2 Na
Quantity 50 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-2-carboxy-2-(3-thienyl)acetyl]amino]-3,3-dimethyl-7-oxo-','sodium salt (1:2)','(2S,5R,6R)-, 3-Thiophenemalonamic acid','N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-','disodium salt','D-(-)- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[(carboxy-3-thienylacetyl)amino]-3,3-dimethyl-7-oxo-','disodium salt','[2S-[2α,5α,6β(S*)]]-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','6-[[(2R)-carboxy-3-thienylacetyl]amino]-3,3-dimethyl-7-oxo-','disodium salt','(2S,5R,6R)- (9CI), 6-D(-)-(α-Carboxy-3-thienylacetamido)penicillanic acid disodium salt, 6-D-(-)-α-Carboxy-3-thienylmethylacetamido penicillanic acid, AB 2288, BRL 2288, Disodium ticarcillin, Monapen, Ticar, Ticarcillin disodium, Ticarcillin disodium salt, Ticarcillin sodium, Ticarcillin sodium salt, Ticarpen, α-Carboxy-3-thienylmethylpenicillin disodium salt, D-Ticarcillin Disodium
InChI Formula InChI=1S/C15H16N2O6S2.2Na/c1-15(2)9(14(22)23)17-11(19)8(12(17)25-15)16-10(18)7(13(20)21)6-3-4-24-5-6;;/h3-5,7-9,12H,1-2H3,(H,16,18)(H,20,21)(H,22,23);;/q;2*+1/p-2/t7-,8-,9+,12-;;/m1../s1
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C(=O)[O-])c3ccsc3)C(=O)N2[C@H]1C(=O)[O-]
IUPAC Name disodium;(2S,5R,6R)-6-[[(2R)-2-carboxylato-2-thiophen-3-ylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 428.39
Formula Weight 428.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.