missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Tick-Borne Encephalitis Virus Proteome Microarray
Description
- Microarray Content: Genome polyprotein of tick-borne encephalitis virus cleaved into 13 chains: peptide 2k, capsid protein C, prM, peptide pr, small envelope protein M, envelope protein E, non-structural protein 1, non-structural protein 2A, serine protease subunit NS2B, serine protease NS3, non-structural protein 4A, non-structural protein 4B, RNA-directed RNA polymerase NS5
- Peptide Length/Overlap: 15 aa/ 14 aa
- Number of Peptides: 3414 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
Specifications
| Coating | PEGMA/PMMA |
| Description | 1 array copy per microarray |
| Quantity | 1 Slide |
| Method of Attachment | Combinatorial peptide synthesis by amino acid laser printing |
| Thickness (Metric) | 1 mm |
| Certifications/Compliance | ISO 9001:2015 |
| Color | Transparent |
| Regulatory Status | RUO |
| Barcode | Individual barcode for each microarray |
| Material | Microscopic Glass |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?