missing translation for 'onlineSavingsMsg'
Learn More

Tiglic acid, MedChemExpress

Product Code. 16469366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469366 Supplier Medchem Express Supplier No. HYW012999
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Tiglic acid is a monocarboxylic unsaturated organic acid found in croton oil and in several other natural products. Tiglic aci has a role as a plant metabolite.

Tiglic acid is a monocarboxylic unsaturated organic acid found in croton oil and in several other natural products. Tiglic aci has a role as a plant metabolite.

Specifications

Chemical Name or Material Tiglic acid
CAS 80-59-1
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C5H8O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (998.80 mM; Need ultrasonic) ∣H2O : 7.69 mg/mL (76.81 mM; Need ultrasonic)
SMILES C/C=C(C)/C(O)=O
Molecular Weight (g/mol) 100.12
Formula Weight 100.12
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.