missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tigloyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Tigloyl L-Carnitine-D3 (N-methyl-D3) Chloride |
| Molecular Formula | C12 D3 H19 N O4 . Cl |
| Quantity | 0.005 g |
| Synonym | Tiglylcarnitine-d3 hydrochloride, Tigloyl-L-carnitine-d3 (N-methyl-d3) hydrochloride, Tigloyl L-carnitine-D3 (N-methyl-D3) chloride, (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[[(2 E)-2-methyl-1-oxo-2-buten-1-yl]oxy]-1-propanaminium chloride |
| InChI Formula | InChI=1 S/C12H21NO4.ClH/c1-6-9(2)12(16)17-10(7-11(14)15)8-13(3,4)5;/h6,10 H,7-8H2,1-5H3;1 H/b9-6+;/t10-;/m1./s1/i3D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)\C(=C\C)\C |
| IUPAC Name | [(2 R)-3-carboxy-2-[(E)-2-methylbut-2-enoyl]oxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 282.779 |
| Formula Weight | 282.143 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction