Learn More
Tiki1/TRABD2A Mouse anti-Human, Clone: 901705, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB8376-SP
Encompass
Product Code. 15780015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Tiki1/TRABD2A Monoclonal specifically detects Tiki1/TRABD2A in Human samples. It is validated for Western Blot.