Learn More
BOLA Tiles
Product Code. 10640562
Click to view available options
Dimensions (L x W):
300 x 2 mm
300 x 3 mm
300 x 4 mm
300 x 5 mm
300 x 6 mm
300 x 8 mm
300 x 10 mm
300 x 15 mm
Unit Size:
Each
Product Code. 10640562
Brand: BOLA S180512
Only null left
Add to basket
Add to basket
Tiles virgin PTFE
Standard sizes with different thicknesses.
Specifications
Absorbancy Non-absorbent
Dimensions (L x W) 300 x 8 mm