missing translation for 'onlineSavingsMsg'
Learn More

Tin, 99.8%, powder, -325 mesh

Product Code. 10125330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10125330 100 g Plastic bottle 100g
10371324 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10125330 Supplier Thermo Scientific Acros Supplier No. 317801000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Gray
Melting Point 231.9°C
Boiling Point 2270.0°C
Density 7.3000g/mL
Quantity 100 g
Packaging Plastic bottle
Assay Percent Range 99.8%
Form Powder
Linear Formula Sn
Fieser 01,1168; 02,414; 11,519; 12,486; 13,298; 17,333
Merck Index 15, 9603
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids and hot potassium hydroxide, solutions
Formula Weight 118.69
Percent Purity 99.8%
Chemical Name or Material Tin, -325 mesh
Show More Show Less
Product Identifier
  • Tin, powder, -325 mesh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.