missing translation for 'onlineSavingsMsg'
Learn More

Tin Ditriflate, TRC

Product Code. 30454003
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454003 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454003 Supplier Toronto Research Chemicals Supplier No. TRCT44410310MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tin Ditriflate
CAS 62086-04-8
Molecular Formula C2O6F6S2Sn
Quantity 10 mg
Synonym Tin(2+) Triflate, Tin(II) Triflate, Tin(II) Trifluoromethanesulfonate, Trifluoromethanesulfonic Acid Tin(2+) Salt, Stannous Triflate, Stannous Trifluoromethanesulfonate, Stannous Trifluoromethylsulfonate, Tin Bis(trifluoromethanesulfonate)
InChI Formula InChI=1S/2CHF3O3S.Sn/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2
SMILES [Sn+2].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
IUPAC Name tin(2+);trifluoromethanesulfonate
Molecular Weight (g/mol) 416.84
Formula Weight 417.8062
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.