missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 0.025mm (0.001in) thick, hard, 99.9% (metals basis)

Product Code. 11336887
Change view
Click to view available options
Quantity:
15 x 15 cm
15 x 75 cm
15 x 300 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336887 15 x 15 cm Each
11346887 15 x 75 cm Each
11356887 15 x 300 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11336887 Supplier Thermo Scientific Alfa Aesar Supplier No. 010789.VA
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Form: Approx. 4.2g/15 x 15cm; Temper: as rolled | CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Color Silver to Gray
Appearance Silver Gray foil
Quantity 15 x 15 cm
Odor Odorless
Assay Percent Range 99.9% (Metals basis)
Form Foil
Chemical Name or Material Tin, Hard

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.