missing translation for 'onlineSavingsMsg'
Learn More

Tin foil, 0.1mm (0.004in) thick, 98.8% (metals basis)

Product Code. 15436585
Change view
Click to view available options
Quantity:
30 x 150 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436585 30 x 150 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436585 Supplier Thermo Scientific Alfa Aesar Supplier No. 044154.RG
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-31-5
Quantity 30 x 150 cm
Odor Odorless
Weight ≈329g/30x150cm
Form Foil
Molecular Formula Sn
MDL Number MFCD00133862
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES [Sn]
IUPAC Name tin
Molecular Weight (g/mol) 118.71
PubChem CID 5352426
ChEBI CHEBI:32990
Percent Purity 98.8%
Assay (metals basis)
Chemical Name or Material Tin foil, 0.1mm (0.004 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.