missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, 98+%

Product Code. 10731881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10731881 500 g Plastic bottle 500g
10022840 100 g Plastic bottle 100g
10053623 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10731881 Supplier Thermo Scientific Acros Supplier No. 222665000
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
Molecular Weight (g/mol) 225.63
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate

Spécification

Boiling Point 652.0°C
Melting Point 37.0°C to 38.0°C
Color White
Physical Form Adhering Crystalline Powder or Crystals and Chunks
Quantity 500 g
Packaging Plastic bottle
Assay Percent Range 98.0 to 103% (Iodimetry)
Density 2.7100g/mL
Linear Formula SnCl2·2H2O
Fieser 01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68
Merck Index 15, 8910
Solubility Information Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
Formula Weight 225.63
Percent Purity ≥98%
Chemical Name or Material Tin(II) chloride dihydrate, p.a.
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Tin(II) chloride dihydrate
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.