Learn More
Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis)
Description
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 43°C to 46°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Linear Formula | SnCl2·xH2O |
| UN Number | UN3260 |
| Merck Index | 14,8783 |
| Solubility Information | Soluble in ethanol,acetone,ether and tetrahydrofuran. Insoluble in xylene. |
| IUPAC Name | dichlorotin |
| Formula Weight | 189.62 (Anhydrous) |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.