missing translation for 'onlineSavingsMsg'
Learn More

Tin (II) Chloride, TRC

Product Code. 30450207
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30450207 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30450207

Brand: Toronto Research Chemicals TRCT44380010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stannous Chloride
CAS 7772-99-8
Molecular Formula SnCl2
Quantity 10 g
Synonym Tin Dichloride, Dichlorotin, Tin Protochlorie, Tin Chloride, Tin (II) Chloride, Stannous Dichloride
InChI Formula InChI=1S/2ClH.Sn/h2*1H;/q;;+2/p-2
SMILES Cl[Sn]Cl
IUPAC Name dichlorotin
Molecular Weight (g/mol) 189.6
Formula Weight 189.8399
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.