missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) oxalate, 98%

Product Code. 11449193
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. Quantity unitSize
11449193 2500 g 2500g
11429193 100 g 100g
11439193 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11449193

Brand: Thermo Scientific Alfa Aesar A11689.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 814-94-8 | C2O4Sn | 206.728 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-94-8
Molecular Formula C2O4Sn
Molecular Weight (g/mol) 206.728
MDL Number MFCD00040678
InChI Key OQBLGYCUQGDOOR-UHFFFAOYSA-L
Synonym stannous oxalate, tin ii oxalate, tin oxalate, tin 2+ oxalate, ethanedioic acid, tin 2+ salt 1:1, stavelan cinaty czech, unii-sar72fe8eh, sar72fe8eh, stavelan cinaty
PubChem CID 13149
IUPAC Name oxalate;tin(2+)
SMILES C(=O)(C(=O)[O-])[O-].[Sn+2]

Specifications

Melting Point 280°C (decomposition)
Quantity 2500 g
Linear Formula SnC2O4
UN Number UN3288
Beilstein 3708588
Merck Index 14,8786
Formula Weight 206.72
Percent Purity 98%
Density 3.56
Chemical Name or Material Tin (II) oxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.