missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) pyrophosphate

Product Code. 11413830
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11413830

Brand: Thermo Scientific Alfa Aesar 089785.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15578-26-4 | O7P2Sn2 | 411.361 g/mol

Tin(II) pyrophosphate is used to inhibit the formation of volatile sulfur compounds. Further, it is used for cyanide-free tin coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15578-26-4
Molecular Formula O7P2Sn2
Molecular Weight (g/mol) 411.361
MDL Number MFCD00049544
InChI Key GEZAUFNYMZVOFV-UHFFFAOYSA-J
Synonym stannous pyrophosphate, technescan pyp, ditin pyrophosphate, tin ii pyrophosphate, tin 2+ pyrophosphate, unii-4dnt29ec86, stannous pyrophosphate usan, tin 2+ diphosphate 1:2, pyro stannous phosphate, ditin 2+ pyrophosphate 4-
PubChem CID 66379
IUPAC Name phosphonato phosphate;tin(2+)
SMILES [O-]P(=O)([O-])OP(=O)([O-])[O-].[Sn+2].[Sn+2]

Specifications

Melting Point 400°C (decomposition)
Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 411.32
Density 4.009
Chemical Name or Material Tin(II) Pyrophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.