missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) trifluoromethanesulfonate, 98%

Product Code. 10717353 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10717353 1 g Plastic bottle 1g
10792944 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10717353 Supplier Thermo Scientific Acros Supplier No. 312530010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62086-04-8 | C2F6O6S2Sn | 416.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62086-04-8
Molecular Formula C2F6O6S2Sn
Molecular Weight (g/mol) 416.85
MDL Number MFCD00191251
InChI Key IACSYIHNPIQPMP-UHFFFAOYSA-M
Synonym tin ii trifluoromethanesulfonate
PubChem CID 131676109
IUPAC Name molecular hydrogen;tin(2+);trifluoromethanesulfonate
SMILES [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]

Specifications

Melting Point >300.0°C
Color White to Yellow
Physical Form Powder
Quantity 1 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 97.5% min. (ex Sn)
Solubility Information Solubility in water: soluble
Formula Weight 416.85
Percent Purity 98%
Chemical Name or Material Tin(II) trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Tin(II) trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.