missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride hydrate, 98%

Product Code. 11347307
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
11347307 500 g 500g
11337307 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11347307

Brand: Thermo Scientific Alfa Aesar 011570.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-06-9 | Cl4Sn | 260.51 g/mol

Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-06-9
Molecular Formula Cl4Sn
Molecular Weight (g/mol) 260.51
MDL Number MFCD00149864
InChI Key HPGGPRDJHPYFRM-UHFFFAOYSA-J
Synonym tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4
PubChem CID 24287
IUPAC Name tetrachlorostannane
SMILES Cl[Sn](Cl)(Cl)Cl

Specifications

Melting Point 56°C
Physical Form Lump
Quantity 500 g
Assay Percent Range 98%
Solubility Information Soluble in water and alcohol.
Formula Weight 260.50 (anhydrous)
Chemical Name or Material Tin(IV) chloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.