missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) Chloride Pentahydrate, 98%, Honeywell Fluka™

Product Code. p-7120874 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
5g
Product Code. Quantity Packaging unitSize
15632660 5 g Poly Bottle 5g
15612660 100 g Poly Bottle 100g
15622660 1 kg Poly Bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 15632660

Brand: Honeywell-Fluka 2446785G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-06-9
Molecular Formula Cl4H10O5Sn
Molecular Weight (g/mol) 350.59
MDL Number MFCD00149864
InChI Key KHMOASUYFVRATF-UHFFFAOYSA-J
Synonym tin iv chloride pentahydrate, tetrachlorostannane pentahydrate, tin chloride pentahydrate, stannic chloride pentahydrate, stannane, tetrachloro-, pentahydrate, ccris 6330, tin iv chloride, pentahydrate 1:4:5, cl4sn.5h2o, stannic chloride 5h2o
PubChem CID 61442
IUPAC Name tin(4+) pentahydrate tetrachloride
SMILES O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

Specifications

Color White to Yellow
Quantity 5 g
Linear Formula SnCl4 · 5H2O
UN Number UN2440
Formula Weight 350.6g/mol
Percent Purity 98%
Packaging Poly Bottle
Appearance Powder or Chunks
Chemical Name or Material Tin(IV) Chloride Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.