missing translation for 'onlineSavingsMsg'
Learn More

Tin wire, 0.5mm (0.02in) dia, Puratronic™, 99.9985% (metals basis)

Product Code. 11316368
Change view
Click to view available options
Quantity:
1 m
5 m
25 m
Unit Size:
1m
25m
5m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11316368 5 m 5m
11396358 1 m 1m
11306368 25 m 25m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11316368 Supplier Thermo Scientific Alfa Aesar Supplier No. 011906.G5
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-31-5 | Sn | 118.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-31-5
Molecular Formula Sn
Molecular Weight (g/mol) 118.71
MDL Number MFCD00133862
InChI Key ATJFFYVFTNAWJD-UHFFFAOYSA-N
Synonym powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
PubChem CID 5352426
ChEBI CHEBI:32990
IUPAC Name tin
SMILES [Sn]

Specifications

Quantity 5 m
Odor Odorless
Weight ≈1.43g/m
Form Wire
Percent Purity 99.9985%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tin wire, 0.5mm (0.02 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.