missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Bromophenol Blue Solution

For use with Lovibond test kits, colorimeters and spectrophotometers

Brand:  Lovibond™ 451621

Code : NEW

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15312025

  • 32.40 € / 100mL
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

  • Bromophenol Blue Solution
  • pH Indicator
  • 100mL and 250mL Bottles
Specifications

Specifications

pH Test
For use with Lovibond test kits, colorimeters and spectrophotometers
100 mL
Bromophenol Blue Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers