missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Chromium Hexavalent Powder Pack Reagent, 100 tests

Product Code. 15321705
Change view
Click to view available options
Range:
0.005 to 0.5 mg/L, 0.02 to 2.0 mg/L
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
15321705 0.005 to 0.5 mg/L, 0.02 to 2.0 mg/L Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15321705 Supplier Lovibond™ Supplier No. 537310
  • ONLINE EXCLUSIVE PRICE
    35.60 €
    27.60 € / Pack of 100
    Save 8.00 €
    22% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated for use in Lovibond test kits, colorimeters and spectrophotometers.

  • Range: 0.005 to 0.5mg/L and 0.02 to 2.0mg/L
  • For use in our test kits, colorimeters and spectrophotometers
  • Pre-measured, unit dose reagents
  • Easy to handle, easy to open and easy to pour
  • Powder pack reagents have a 5 year shelf life
  • Case of 100

Specifications

For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
No. of Tests Case of 100
Range 0.005 to 0.5 mg/L, 0.02 to 2.0 mg/L
Type Chromium Hexavalent Powder Pack Reagent
compliance-icons compliance-icons
Product Identifier
  • chromium hexavalent reagent powder
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sulfamic acid, cetrimonium salts
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.