missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Iron Low Range Tablets

Product Code. 15321835
Change view
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15321835 250 Tablets Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15321835 Supplier Lovibond™ Supplier No. 515371BT
  • ONLINE EXCLUSIVE PRICE
    200.00 €
    197.80 € / Pack of 250
    Save 2.20 €
    1% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits

  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents

Specifications

For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type Iron Low Range Tablets
compliance-icons
Product Identifier
  • X250 IRON LR TABLETS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: adipic acid, calciumthioglycolate trihydrate, disodium 4,4'-[3-(pyridin-2-yl)-1,2,4-triazine-5,6-di
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.