missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Total Alkalinity M Tablets

Product Code. 15311825
Click to view available options
Quantity:
250 Tablets
Unit Size:
Pack of 250
This item is not returnable. View return policy

Product Code. 15311825

Brand: Lovibond™ 515321BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits

  • Alkalinity Total M Tablets
  • Method: Tablet Count
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents

Specifications

For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type Alkalinity Total M Tablets
compliance-icons
Product Identifier
  • X250 TOTAL ALKALINITY M/2 TABLETS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium bisulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.