missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tioconazole, 98+%

Product Code. 15463229
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463229 5 g 5g
15453229 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463229 Supplier Thermo Scientific Chemicals Supplier No. J60459.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent inhibitor of cytochrome-P450 | CAS: 65899-73-2 | C16H13Cl3N2OS

Potent inhibitor of cytochrome-P450

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65899-73-2
Molecular Formula C16H13Cl3N2OS
Synonym Thioconazole

Specifications

Quantity 5 g
Description White to Off-white
Format Powder
For Use With (Application) Potent inhibitor of cytochrome-P450
Formula Weight 387.71g/mol
Product Type Signal Transduction Reagent
Form Powder
Assay ≥98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.