missing translation for 'onlineSavingsMsg'
Learn More

Tipifarnib, MedChemExpress

Product Code. 16485006
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485006 100 mg 100mg
16450367 50 mg 50mg
16426727 5 mg 5mg
16404018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485006 Supplier Medchem Express Supplier No. HY10502
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tipifarnib (IND 58359) binds to and inhibits farnesyltransferase (FTase) with an IC50 of 0.86 nM. Antineoplastic activity.

Tipifarnib (IND 58359) binds to and inhibits farnesyltransferase (FTase) with an IC50 of 0.86 nM. Antineoplastic activity.

Specifications

Chemical Name or Material Tipifarnib
CAS 192185-72-1
Color White
Purity Grade Notes Research
Molecular Formula C27H22Cl2N4O
Quantity 100 mg
Synonym IND 58359 R115777
Solubility Information DMSO : 14.29 mg/mL (29.20 mM; Need ultrasonic)
SMILES O=C1N(C2=C(C(C3=CC=CC(Cl)=C3)=C1)C=C(C=C2)[C@@](N)(C4=CN=CN4C)C5=CC=C(C=C5)Cl)C
Molecular Weight (g/mol) 489.4
Formula Weight 489.4
Percent Purity 96.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.