missing translation for 'onlineSavingsMsg'
Learn More

Tiplaxtinin, MedChemExpress

Product Code. 16416696
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16416696 100 mg 100mg
16486398 10 mg 10mg
16472507 50 mg 50mg
16468987 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16416696 Supplier Medchem Express Supplier No. HY15253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tiplaxtinin is a selective and orally efficacious inhibitor of plasminogen activator inhibitor-1 (PAI-1) with IC50 of 2.7 μM.

Tiplaxtinin is a selective and orally efficacious inhibitor of plasminogen activator inhibitor-1 (PAI-1) with IC50 of 2.7 μM.

Specifications

Chemical Name or Material Tiplaxtinin
CAS 393105-53-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H16F3NO4
Quantity 100 mg
Synonym PAI-039 Tiplasinin
Solubility Information DMSO : 100 mg/mL (227.59 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C(O)=O)C1=CN(CC2=CC=CC=C2)C(C1=C3)=CC=C3C4=CC=C(OC(F)(F)F)C=C4
Molecular Weight (g/mol) 439.38
Formula Weight 439.38
Percent Purity 98.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.