missing translation for 'onlineSavingsMsg'
Learn More

TIQ-A, TRC

Product Code. 30451219
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30451219 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451219 Supplier Toronto Research Chemicals Supplier No. TRCT4451001MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 420849-22-5
Molecular Formula C11 H7 N O S
Quantity 1 mg
InChI Formula InChI=1S/C11H7NOS/c13-10-8-4-2-1-3-7(8)9-5-6-14-11(9)12-10/h1-6H,(H,12,13)
SMILES O=C1Nc2sccc2c3ccccc13
IUPAC Name 4H-thieno[2,3-c]isoquinolin-5-one
Molecular Weight (g/mol) 201.24
Formula Weight 201.0248
Chemical Name or Material TIQ-A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.