missing translation for 'onlineSavingsMsg'
Learn More

Tiron, Indicator, Solstice

Product Code. 15617390 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15617390 100 g Plastic bottle 100g
15627390 10 g Plastic bottle 10g
15637390 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15617390 Supplier Honeywell-Fluka Supplier No. 89460100G
  • ONLINE EXCLUSIVE PRICE
    236.00 €
    198.34 € / 100g
    Save 37.66 €
    15% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator (for complexometry)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270573-71-2
Molecular Formula C6H6Na2O9S2
Molecular Weight (g/mol) 332.205
MDL Number MFCD00149531
InChI Key ZLRROLLKQDRDPI-UHFFFAOYSA-L
PubChem CID 2723960
IUPAC Name disodium;4,5-dihydroxybenzene-1,3-disulfonate;hydrate
SMILES C1=C(C=C(C(=C1S(=O)(=O)[O-])O)O)S(=O)(=O)[O-].O.[Na+].[Na+]

Specifications

Melting Point 300°C
Packaging Plastic bottle
Linear Formula (OH)2C6H2(SO3Na)2 · H2O
UN Number NONH for all modes of transport
Color Violet to White
Solubility Information 1350g/L
pH 5.0
Quantity 100 g
Physical Form Solid
Formula Weight 332.22g/mol
Percent Purity min. 95.0%
Chemical Name or Material Tiron
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.