missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Chloride, ReagentPlus™, 99.9% trace metals basis, Honeywell™

Product Code. 15651840 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
200 g
Packaging:
Glass Bottle
Unit Size:
200g
This item is not returnable. View return policy

Product Code. 15651840

Brand: Honeywell Research Chemicals 208566200G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99.9% trace metals basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 136.4°C
Melting Point -24°C
CAS 7550-45-0
Color Colorless to Yellow
Physical Form Liquid
Quantity 200 g
Molecular Formula Cl4Ti
Linear Formula TiCl4
MDL Number MFCD00011267
UN Number UN1838
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
IUPAC Name titanium(4+);tetrachloride
Molecular Weight (g/mol) 189.667
PubChem CID 160960
Formula Weight 189.68g/mol
Percent Purity 99.9% (trace metals basis)
Grade ReagentPlus
Packaging Glass Bottle
Vapor Pressure 9.6mmHg (20°C), 50mmHg (55°C)
Density 1.73g/mL at 20°C
Chemical Name or Material Titanium(IV) Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.