missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Dioxide, Extra Pure, SLR, Fisher Chemical

Product Code. 10695502 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10695502 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10695502 Supplier Fisher Chemical Supplier No. T/1900/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 2900°C
Melting Point 1855°C
CAS 13463-67-7
Color White
pH 3.5 to 4.5
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula O2Ti
MDL Number MFCD00011269 MFCD00210650
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILES O=[Ti]=O
IUPAC Name dioxotitanium
Molecular Weight (g/mol) 79.87
PubChem CID 26042
ChEBI CHEBI:32234
Formula Weight 79.88g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.