missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) Isopropoxide, TRC

Product Code. 30448958
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30448958 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30448958 Supplier Toronto Research Chemicals Supplier No. TRCT4468001G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium(IV) Isopropoxide
CAS 546-68-9
Molecular Formula C12 H28 O4 Ti
Quantity 1 g
Synonym Isopropyl Alcohol Titanium(4+) Salt Titanium Isopropoxide (Ti(OC3H7)4) (7CI), 5N, 5N (titanate), A 1, A 1 (titanate), AKT 872, Bistrater H-NDH 510C, Isopropyl Orthotitanate, Isopropyl Titanate(IV) ((C3H7O)4Ti), NDH 510C, Orgatix TA 10, TA 10, TIPT, TPT, TPTA 1, Tetraisopropanolatotitanium, Tetraisopropoxytitanium, Tetraisopropoxytitanium(IV), Tetraisopropyl Orthotitanate, Tetraisopropyl Titanate, Tetrakis(isopropanolato)titanium, Tetrakis(isopropoxy)titanium, Tetrakis(isopropylato)titanium(IV), Tetrakis(isopropyloxy)titanium, Tilcom TIPT, Titaium tetraisopropoxide, Titanium Isopropoxide, Titanium Isopropylate, Titanium Tetraisopropoxide, Titanium Tetraisopropylate, Titanium Tetrakis(iso-propoxide), Titanium Tetrakis(isopropoxide), Titanium(4+) Isopropoxide, Titanium(IV) isopropoxide, Tetrakis(1-methylethoxy)titanium, Titanum-(IV)-isopropoxide, Tyzor TPT, Vertec RJCE, Vertec TIPT, Vertec XI 900, Vertec XL 110
InChI Formula InChI=1S/4C3H7O.Ti/c4*1-3(2)4;/h4*3H,1-2H3;/q4*-1;+4
SMILES CC(C)O[Ti](OC(C)C)(OC(C)C)OC(C)C
IUPAC Name tetra(propan-2-yloxy)titanium
Molecular Weight (g/mol) 284.22
Formula Weight 284.1467
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.