missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 99+%

Product Code. 15482957
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
Product Code. Quantity unitSize
15482957 500 g 500g
15462957 10 g 10g
15472957 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 15482957

Brand: Thermo Scientific Alfa Aesar 077124.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5593-70-4 | C16H40O4Ti | 344.359 g/mol

Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5593-70-4
Molecular Formula C16H40O4Ti
Molecular Weight (g/mol) 344.359
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Specifications

Boiling Point 310°C to 314°C
Melting Point -55°C
Color Yellow
Physical Form Liquid
Quantity 500 g
Linear Formula Ti[O(CH2)3CH3]4
UN Number UN2924
Beilstein 4148236
Flash Point 41°C (170°F)
Solubility Information Miscible with aliphatic,aromatic,chlorinated and oxygenated solvents.
Formula Weight 340.36
Percent Purity ≥99%
Odor Alcohol-like
Refractive Index 1.49
Sensitivity Moisture sensitive
Density 0.998 g/mL
Chemical Name or Material Titanium (IV) n-butoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) n-butoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Butyl titanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.