missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Solstice

Product Code. 15661390
Change view
Click to view available options
Quantity:
250 g
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
250g
25kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661390 6 x 1 kg Pack of 6
15641390 250 g 250g
15631390 1 kg 1kg
15651390 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15661390 Supplier Honeywell-Fluka Supplier No. 140276X1KG
  • ONLINE EXCLUSIVE PRICE
    510.00 €
    460.63 € / Pack of 6
    Save 49.37 €
    9% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Powder
Quantity 6 x 1 kg
Density 4.26g/mL at 25°C
Linear Formula TiO2
UN Number NONH for all modes of transport
Formula Weight 79.87
Percent Purity 99.0 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Titanium(Iv) Oxide, Meets analytical specification of Ph. Eur.BP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.