missing translation for 'onlineSavingsMsg'
Learn More

Titanium(Iv) Oxide, ReagentPlus™, Honeywell™

Product Code. 15601390 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15601390 1 kg 1kg
15611390 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15601390 Supplier Honeywell Research Chemicals Supplier No. 140211KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Quantity 1 kg
Linear Formula TiO2
UN Number NONH for all modes of transport
Formula Weight 79.87g/mol
Percent Purity ≥99%
Grade ReagentPlus
Density 4.26g/mL at 25°C
Chemical Name or Material Titanium(Iv) Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.